Corning® 3D Tissue Clearing Reagent, Corning
Supplier: Corning
Corning®
5733 5731 5732 5731 5732 5733
CA76463-342CS 205.87 CAD
76423-682 76423-684 CA76463-342 76423-680 CA76463-344 CA76463-346
Corning® 3D Tissue Clearing Reagent, Corning
Cell Culture Supplements and Additives
Corning 3D Clear tissue clearing designed for imaging 3D CC models and high-throughput processing.
  • Rapid tissue clearing
  • Easy to Use
  • No Special Equipment Required
  • Compatible with IF, FP and other Fluorescent lables
  • Microplate and Automation Compatible
  • Reversible to Follow up with 2D H&E/IHC
The Corning 3D Clear tissue clearing technique is designed specifically to support imaging for 3D cell culture models and plate-based high-throughput processing. When paired with fluorescent labeling (e.g. fluorescent protein, immunofluorescence, chemical dyes) and high content confocal microscopy, Corning 3D Clear Reagent allows for complete 3D cell culture model characterization and more accurate drug screening.
Refractive index of 1.48
Caution: Not intended for use in diagnostic or therapeutic procedures.