Corning® 3D Clear Starter Kit, 3D Clear Reagent, Antibody, Blocking, Penetration and Washing Buffers, Corning
Supplier: Corning
Corning®
5730 5730
CA76463-340CS 983.44 CAD
CA76463-340 76423-678
Corning® 3D Clear Starter Kit, 3D Clear Reagent, Antibody, Blocking, Penetration and Washing Buffers, Corning
Cell Culture Supplements and Additives
Starter Kit combines Corning 3D Clear with buffer solutions required to process 3D tissue cultures.
  • Rapid tissue clearing
  • Easy to Use
  • No Special Equipment Required
  • Compatible with IF, FP and other Fluorescent lables
  • Microplate and Automation Compatible
  • Reversible to Follow up with 2D H&E/IHC
The Starter Kit combines 30 ml of Corning 3D Clear with the four buffer solutions required to process 3D tissue cultures for immunolabeling.  All you need to do is add your immunolabels and nuclear stains. This kit comes with enough material to process three 96-well plates of 3D cell culture models.
Includes: Corning 3D Clear reagent (30 ml), Corning 3D Clear antibody buffer (30 ml), Corning 3D Clear blocking buffer (30 ml), Corning 3D Clear penetration buffer (30 ml), Corning 3D Clear washing buffer 10X (70 ml)
Caution: For best results, refrigerate buffer after opening.