BIOne Single-Use Bioreactor Systems, Distek
Supplier: DISTEK, INC.
2022-1001 2022-1002 2022-1014 2022-1015 2022-1018 2022-1024 2022-1025 2022-1028 2022-1029 2022-1005 2022-1006 2022-1016 2022-1017 2022-1019 2022-1023 2022-1026 2022-1027 2022-1030 2022-1031
76419-906EA 1211.86 CAD
76419-906 76419-908 76419-910 76419-912 76419-914 76419-918 76419-920 76419-922 76419-924 76419-930 76419-932 76419-934 76419-936 76419-938 76419-940 76419-942 76419-944 76419-946 76419-948 76419-950
BIOne Single-Use Bioreactor Systems, Distek
Bioreactors
Distek, Inc. has developed a benchtop scale single-use bioreactor (SUB) for mammalian cell growth, cell therapy, insect cells, and other unique applications performed in a stirred tank bioreactor. The gamma irradiated BIOne SUB is engineered with a disposable headplate welded to a triple-layered liner that can be easily inserted into a non sterile bioreactor glass vessel, converting it to a disposable SUB within a matter of seconds.
  • Customizable above and below the headplate
  • 2 and 5 L working volume
  • Optional single-use pH probe
  • Increased throughput
More Product Information