BIOne Autoclavable Bioreactor Assemblies for Microbial Applications, Distek
Supplier: DISTEK, INC.
2022-1506 2022-1508 2022-1510
76419-928EA 17969.34 CAD
76419-928 76419-954 76419-956
BIOne Autoclavable Bioreactor Assemblies for Microbial Applications, Distek
Bioreactors
The BIOne autoclabable bioreactor is available in a range of sizes including; 2, 5 and 10 L working volume. All metal components are manufactured from 316L stainless steel with electropolish.
The standard microbial configuration includes: glass vessel, stainless steel headplate, vessel stand, dual rushton impellers, baffles, cooling finger, sparger, triple addition port, overlay gas inlet port, exhaust condenser, harvest tube, thermowell, sample down tube, sensor ports, foam probe, septum port, addition ports and port plugs.
Ordering information: If for use with a controller other than the BIOne 1250, a BIOne motor adapter assembly is required. Motor adapters are controller motor specific. For more details please contact your local Avantor sales office.