BIOne Autoclavable Bioreactor Assemblies for Cell Culture Applications, Distek
Supplier: DISTEK, INC.
2022-1505 2022-1507 2022-1509 2800-0511 2800-0512 2800-0513 2800-0525 2800-0527 2800-0530 2800-0610 2800-0627 2800-0635 2800-0922 2800-0962
76419-926EA 13696.18 CAD
76419-926 76419-952 76419-958 76420-004 76420-006 76420-008 76420-010 76420-012 76420-014 76420-016 76420-018 76420-020 76420-022 76420-024
BIOne Autoclavable Bioreactor Assemblies for Cell Culture Applications, Distek
Bioreactors
The BIOne autoclabable bioreactor is available in a range of sizes including; 2, 5 and 10 L working volume. All metal components are manufactured from 316L stainless steel with electropolish.
The standard cell culture configuration includes: glass vessel, stainless steel headplate, vessel stand, dual pitched blade impellers, micro sparger, triple addition port, overlay gas inlet port, exhaust gas outlet port, harvest tube, thermowell, sample downtube, sensor ports, single addition ports, foam probe and port plugs.
Ordering information: If for use with a controller other than the BIOne 1More Product Information