SP Bel-Art® Sterileware® Powder Lance Samplers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F36974-0420 F36974-0600 F36974-0950
76402-892PK 329.2 CAD
76402-892 76402-894 76402-896
SP Bel-Art® Sterileware® Powder Lance Samplers, Bel-Art Products, a part of SP
Samplers Solid Samplers
A simple insert, rotate, and withdraw motion allows cross sectional sampling of non-flowing, bulk powders.
  • Open blade works well with sticky, cohesive powders
  • Rounded edges help hold sample in place
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) lance is heat sealed in individual polyethylene bag
  • Gamma irradiated sterile (SAL 10⁻⁶)
  • Ready for one time use and disposal
  • Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Available in three lengths.
More Product Information