SP Bel-Art Sterileware® Flat Bottom Sampling Scoops, White, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F36911-0025 F36911-0050 F36911-0250 F36911-0500 F36911-1000
76402-874PK 410.44 CAD
76402-874 76402-876 76402-878 76402-882 76402-884 76402-886
SP Bel-Art Sterileware® Flat Bottom Sampling Scoops, White, Bel-Art Products, a part of SP
Scoops
These sterile scoops are used by pharmaceutical, food, cosmetic and chemical companies for general production or laboratory work.
  • Handle bump acts as thumb rest to help keep the hand away from the scoop; reduces contamination risks
  • Flat bottom allows for use as a weigh boat
  • Approximate volumes can be measured by removing excess powder
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High impact polystyrene (HIPS) scoops heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10⁻⁶)
  • Ready for one time use and disposal
  • Lot stamped for accurate tracing; shipped with a certificate of processing for sterilization
These sampling scoops are designed for retrieving samples from a larger source. Capacity/volumes are estimated and are not meant for taking exact sample weights/volumes.
Ordering information: Sterilized products are not returnable.