SP Bel-Art Sterileware® Pharma Scoops, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F36909-0030 F36909-0060 F36909-0125 F36909-0250 F36909-0500 F36909-1000 F36909-2500 F36910-0060 F36910-0125 F36910-0500 F36910-1000 F36910-2500
76402-860PK 429.49 CAD
76402-860 76402-862 76402-864 76402-866 76402-868 76402-870 76402-872 76469-934 76469-936 76469-938 76469-940 76469-942 76469-944 76469-946
SP Bel-Art Sterileware® Pharma Scoops, Bel-Art Products, a part of SP
Scoops
Ideal for sampling for both production work and laboratory use, Pharma Scoops are used by pharmaceutical, food, cosmetic, and chemical companies.
  • Bright red color reduces chance of scoop being left behind in a sample; can serve as code to avoid cross contamination; can provide contrasting background for light colored samples
  • High handle location keeps hand further from sample
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High impact polystyrene (HIPS) scoops heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10⁻⁶)
Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization.