Bel-Art Sterileware® Double Bagged Long Handle Sampling Spoons
Supplier: Bel-Art Products, a Part of SP
H36940-1010 H36942-1010 H36944-1010 H36946-1010
76423-428CS 125.53 CAD
76423-428 76423-430 76423-432 76423-434
Bel-Art Sterileware® Double Bagged Long Handle Sampling Spoons
Spoons
Minimizing contamination of small sample collection. Ten individually packaged scoops are sealed in an outer bag to create multiples layers of protection against contaminants and debris.
  • Ideal for use in multi-zone clean rooms
  • FDA grade polystyrene safe for food, drug and cosmetic use
  • Gamma irradiated sterile to SAL 10⁻⁶
  • Lot stamped for accurate lot tracing, and shipped with a certificate of processing for sterilization
The long handle on these spoons enables users to reach into jars, bottles and other confined areas to minimize the potential ofMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable