Ethanol denatured ≥80% (denatured with methanol and ethyl acetate)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
38344-540
CA71007-230CS
2444.07
CAD
CA71007-230
Ethanol denatured ≥80% (denatured with methanol and ethyl acetate)
Ethanol denatured
(DAG-2A)
Formula:
H₃CCH₂OH |
MDL Number:
MFCD00003568 CAS Number: 64-17-5 UN: 1986 ADR: 3 (6.1),II |
Specification Test Results
Ethanol | 80.2% v/v min. |
Appearance (clear, colorless liquid) | Passes test |
Colour (APHA) | 5 max. |
Density at 20°C | 0.8106 g/ml max. |
Acidity (as CH3COOH ) | 0.003% w/v max. |
Ethyl acetate | 0.75 - 0.95% v/v |
Methanol | 12.95 - 14.45% v/v |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...