PolarSafe® Dewar and Traceable® Temperature Data Logger Bundles, Cole Parmer
Supplier: Antyila Scientific
44260-71 44260-72 44260-73 44260-74 44260-75 44260-76
76421-000EA 13859.06 CAD
76421-000 76421-002 76421-004 76421-006 76421-008 76421-010
PolarSafe® Dewar and Traceable® Temperature Data Logger Bundles, Cole Parmer
Dewars and Cryogenic Vessels
The Cole-Parmer® PolarSafe® dewar together with a Traceable® data logger is the perfect combination to safely store your critical samples while maintaining sample integrity.
  • Improve sample protection – maintain specific temperature ranges for extended time periods
  • Maintain sample integrity with Traceable® temperature monitoring
  • Data logger provided with 2-year NIST-traceable calibration certificate
  • Lowest set temperature of –200 °C
  • Durable aluminum construction
The bundle adds on one Traceable® Wi-Fi data logger compatible with TraceableLIVE® cloud service (model 18001-08) designed specifically for temperatures down to –200 °C. Once your data logger is set up, you can securely monitor parameters 24/7, control up to 100 alarms remotely, view your data logging history, run reports in real time, and give access to other team members.