MULTISTIRRER Digital Multiple Positions Magnetic Stirrers,Velp Scientific
Supplier: VELP SCIENTIFIC INC.
F203A0179 F203A0180
76416-214EA 2376.81 CAD
76416-214 76416-216
MULTISTIRRER Digital Multiple Positions Magnetic Stirrers,Velp Scientific
Stirrers Magnetic Stirrers, Multi-position
Digital multi-position magnetic stirrer with 6 or 15 positions, that remains cold even after several days of continuous use.
  • Bright digital display for accurate speed monitoring
  • Exclusive, on-board timer for unattended operation
  • Adjustable reverse operation (time-controlled)
  • Speed regulation from 80 to 1500 rpm
  • SpeedServo™ ensures constant speed even when the viscosity changes
  • Synchronized stirring, same speed ensured
  • Ergonomic - very low height
  • Universal mains voltage/frequency range
Soft start but rapidly reaching set speed, excellent speed control (More Product Information