Metro Horizontal Glove Box Holders for Super Erecta® Wire Shelving and SmartWall™ Shelving, Intermetro Industries
Supplier: Metropolitan Wire
GBHHK4-S GBHHK4-T
76423-254EA 87.55 CAD
76423-254 76423-256
Metro Horizontal Glove Box Holders for Super Erecta® Wire Shelving and SmartWall™ Shelving, Intermetro Industries
Glove Dispensers and Glove Box Holders
Keep gloves conveniently within reach with the horizontal single or triple glove box holder.
  • Easily connects to any Super Erecta® wire shelf or SmartWall™ grid
  • Holds glove boxes up to 3.5×6×10.125"
  • Single or triple box formats available
Built-in Microban® antimicrobial protection inhibits the growth of damaging bacteria, mold, mildew, and fungus that can cause stains and odors. Coating also prevents product degradation. Gloves not included.