Table-Gard Disposable Work Mat, Yellow, Accuform
Supplier: ACCUFORM MANUFACTURING, INC
LPM145X LPM145P LPM145L
76421-950PK 30.51 CAD
76421-950 76421-952 76421-954
Table-Gard Disposable Work Mat, Yellow, Accuform
Surface Protectors Bench Protectors
The self-adhesive mat is repositionable and lifts easily for disposal when work is complete.
  • Provides protection from germs, dirt, and chemical residue
  • Compact and lightweight mat fits in workbags for on-the-go working needs
  • Made of a smooth, thin 2-mil plastic film that is waterproof and tear-resistant
  • Can be discarded in waste or recycle bin (recycle code 5)
  • 12.5×19.5" size creates a workspace large enough for laptops and folios
  • BPA, Latex, and PVC-free material without toxic chemicals
  • The adhesive is safe for indirect food contact (complies with FDA 21 CFR 175.105)
The mat is safe for application to countertops, desktops, conference tables, and restaurant tables.