Anti-Static Cleanroom Tapes with Conductive Adhesive, UltraTape
Supplier: Ultratape Industries
2149GY075P7D 2149GY100P7D
89203-826CS 3104.48 CAD
89203-826 89203-828
Anti-Static Cleanroom Tapes with Conductive Adhesive, UltraTape
Tapes
Particle-free, residue-free tapes.
Specially constructed self adhesive tape that is based on a polyester film that has been coated with an anti-static primer, plus an anti-static pressure sensitive acrylic adhesive layer.