Polyolefin Medium Adhesion Cleanroom Tapes, UltraTape
Supplier: Ultratape Industries
1164WH050P6U 1164WH075P6U 1164WH150P6U
11212-028CS 3679.79 CAD
11212-028 11212-030 11212-034
Polyolefin Medium Adhesion Cleanroom Tapes, UltraTape
Tapes
Particle-free, residue-free tapes.
  • Ultra-low outgassing
  • No residue cleanroom tape
Featuring a polyolefin backing and acrylic adhesive, 1164 is one of the cleanest tapes available in terms of metal content, outgassing, and residue.