Corning® Bottle Top Dispensers, Corning
Supplier: Corning
6840 6841 6842 6843 6844 6845
76420-190CS 622.47 CAD
76420-190 76420-192 76420-194 76420-196 76420-198 76420-200
Corning® Bottle Top Dispensers, Corning
Bottle Top Dispensers
Corning® bottle top dispensers are volumetric instruments designed for convenient and safe dispensing of liquids with no loss of reagent. Depending on the model, they can accommodate volumes from 0.25 to 100 ml.
  • Accurate and reproducible volume setting
  • Fully autoclavable for protection against contamination
  • Rotating valve block for convenient dispensing
  • Convenient 3-position discharge tube for dispensing, sample recovery, and emptying
  • Unique drip-free system for user safety at work
  • Easy in-lab calibration with built-in recalibration mechanism
In-lab calibration tool and set of four thread adapters included with every product.
Certifications: Meets ISO 8655 requirements.
Delivery information: Dispensers up to 10 ml are delivered with adapter sizes: A25, A28, A40, A45. Dispensers up to 25, 50, and 100 ml are delivered with adapter sizes: A25, A28, A38, A45/32.