Captiva / CaptiVac Vacuum Collars and Collection Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
A8961007 A696001000 A8961007B A796 A796G A696001000B
CAA8961007PK 266.02 CAD
CAA8961007 CAA696001000 CAA8961007B CAAGA796 CAAGA796G CAGA696001000B
Captiva / CaptiVac Vacuum Collars and Collection Plates, Agilent Technologies
Seals Microplate Seals
The Captiva vacuum collar forms a seal between the filtration plate and the collection plate.
  • Plates are designed for Captiva filtration and SPEC, Bond Elut 96 applications
  • Plates have regular 1 ml format which offers compatibility with further automation or liquid handling
  • Pierceable cover preserves sample integrity
  • Collar vacuum sealed for maximum efficiency and pre-aligned for trouble-free operation
  • Simple, cost effective solution
Captiva 96-well collection plates are specially designed for use with Captiva filtration plates, SPEC SPE 96-well plates, and Bond Elut 96-well plates. Captiva filtration and SPEC 96-well plates can be joined directly onto Agilent collection plates using a CaptiVac vacuum collar. This forms a preset, pre-aligned seal between the filtration and collection plate. Completely seal the plates using Captiva Pierceable 96-well silicone covers. The silicone covers are specially designed for 96-well autoinjectors, providing easy piercing and removal. This seal positions the outlet tips at a specified distance inside each well, to prevent cross contamination of samples.