VWR® Clinical 4-Way Tube Racks
Supplier: VWR International
76343-404 76343-406 76343-408 76343-410 76343-412
76343-404CS 802.4 CAD
76343-404 76343-406 76343-408 76343-410 76343-412
VWR® Clinical 4-Way Tube Racks
Racks Tube Racks
These autoclavable tube racks have a six-way connection to create various configurations and are designed to hold round, flat and conical blood collection and culture tubes.
  • Uniformly spaced wells help eliminate pipetting errors which enables more accurate results
  • Molded alphanumeric identification for quick location and retrieval of tubes
  • Made from polypropylene that is chemical resistant and easy to clean
  • Racks can be used in water baths and placed in freezer
  • Accommodates all brands of 13- and 17-mm tubes
They can be used as standalone unit or infinite units can be linked to create configurations to fit number of wells and tube sizes needed.
Packaging: Pack of five.