Bond Elut C2 Plates
Supplier: Agilent
Bond Elut
A496111C 7541101C 7541101C A4961150
AG7541101CEA 699.43 CAD
101473-728 AG7541101C 101402-258 101402-256
Bond Elut C2 Plates
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut C2 is a fairly nonpolar sorbent because of the short chain length of the functional group. C2 is often used during the process of method development if analytes are retained too strongly on a C8 or C18 phase.
  • C2 is a low carbon load sorbent
  • Can be used alongside CN and C8 phases
  • Popular for drug extraction from plasma and for flat baselines
The polarity of C2 is slightly lower than a cyano phase for polar interactions.