ICP-MS Nebulizers and Supplies for PerkinElmer, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
8003-0485 8003-0486 8003-0488
AG8003-0485EA 32.15 CAD
AG8003-0485 AG8003-0486 AG8003-0488
ICP-MS Nebulizers and Supplies for PerkinElmer, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Key design features of nebulizers for PerkinElmer ICP-MS systems provide the best precision, efficient drainage, minimal burner blockage, and reduced interferences.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the stand behind warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Many parts in packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
The GemClean Cross-Flow II with GemTips is optimized for ICP-MS (0.009 and 0.013" orifices) and specially manufactured and cleaned for ultratrace analysis. OpalMist PFA, with uptake at 0.4 ml/min, is designed for high-precision analyses requiring high chemical resistance to HF. Alternatively, the MicroMist, micro-uptake nebulizer delivers high performance for small sample volumes.