Bond Elut C18 OH Plates
Supplier: Agilent
Bond Elut
7542703C A3962925 7541601C A496291C A396291C 7541601C A3962950
AG7541601CEA 699.43 CAD
101473-790 101402-254 101402-286 101402-252 101473-740 101473-760 AG7541601C
Bond Elut C18 OH Plates
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut C18 OH is unendcapped, offering tightly controlled silanol activity to support fractionation. C18 OH delivers enhanced retention of polar analytes.
  • Silanol activity permits metabolite fractionation
  • Tight QC tolerances deliver batch-to-batch reproducibility
  • 150 Å pore size expands retention window to higher molecular weight compounds
It is a nonendcapped version of the octadecyl bonded phases that enables the silanols on the silica surface to be more active. This low-load C18 has well-controlled silanol activity that permits the fractionation of metabolites and enhances retention of basic compounds compared to an endcapped C18. The pore size makes it a good choice for peptide extraction.