Nebulizers and Supplies for PerkinElmer
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
8003-0824 8003-0832
AG8003-0824EA 5.29 CAD
AG8003-0824 AG8003-0832
Nebulizers and Supplies for PerkinElmer
Chromatography Supplies Chromatography Spare Parts
These nebulizers for PerkinElmer AA systems provide the best precision, efficient drainage, minimal burner blockage, and reduced interferences.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies.
Order Now
SPECIFICATIONS
Environmentally Preferable