Column Nuts for Shimadzu GC Systems
Supplier: Agilent
8001-0018 8001-0018 8001-0017 8001-0017
AG8001-0018KT 480.23 CAD
103579-252 AG8001-0018 AG8001-0017 103579-958
Column Nuts for Shimadzu GC Systems
Fittings and Connectors
High quality column nuts designed to provide secure fittings in shimadzu GC systems.
  • Agilent column nuts for shimadzu GC systems seamlessly fit with your instrument
One of the most basic tasks in GC, installing the column, can be challenging. The GC oven is a dark, small space, and column installation uses some tiny parts that need to be assembled with precision. Installation of the column into both the inlet and detector must be done properly to get accurate and reproducible results. Agilent offers a selection of capillary column nuts for Shimadzu GC fittings to facilitate good column installation. The Self Tightening column nut, prevents leaking and the need to retighten when using graphite/polyimide blend ferrules recommended for use with oxygen sensitive detectors such as ECD or mass spectrometers.