Epredia™ Thinline Biopsy Cassettes, Epredia
Supplier: Epredia
B851737BE B851737BL B851737GN B851737GY B851737LA B851737LG B851737LI B851737OR B851737PK B851737TN B851737WH B851737YW
76337-868CS 847.86 CAD
76337-868 76337-870 76337-872 76337-874 76337-876 76337-878 76337-880 76337-882 76337-884 76337-886 76337-888 76337-890
Epredia™ Thinline Biopsy Cassettes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
Thinline Biopsy Cassettes are designed for ease of use and print clarity.
  • For biopsy tissue specimens
  • Variety of 12 colors
  • Comes in tubes (sleeves) for PrintMate™ Printer compatibility
  • Includes matching lids attached
  • Pore style: Small squares
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
Cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, whileMore Product Information