Epredia™ TissueLoc™ Cassette IV Biopsy Cassettes, Epredia
Supplier: Epredia
B851750BE B851750BL B851750GN B851750GY B851750LA B851750LG B851750LI B851750OR B851750PK B851750TN B851750WH B851750YW B851925BE B851925BL B851925GN B851925GY B851925LA B851925LG B851925LI B851925OR B851925PK B851925TN B851925WH B851925YW
76337-940CS 585.64 CAD
76337-940 76337-942 76337-944 76337-946 76337-948 76337-950 76337-952 76337-954 76337-956 76337-958 76337-960 76337-962 76337-964 76337-966 76337-968 76337-970 76337-972 76337-974 76337-976 76337-978 76337-980 76337-982 76337-984 76337-986
Epredia™ TissueLoc™ Cassette IV Biopsy Cassettes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
TissueLoc™ Cassette IV biopsy cassettes are designed for ease of use and print clarity.
  • For small biopsy tissue specimens
  • Variety of 12 colors
  • Comes either in plastic tubes (sleeves) or loose bulk packaging
  • Includes matching lids
  • Large lid tab for easy opening
  • Pore style: Small squares
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
Cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closingMore Product Information