Ultra Inert Press Fit Column Connectors, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5190-6979 5190-6980
CA5190-6979PK 230.12 CAD
CA5190-6979 CA5190-6980
Ultra Inert Press Fit Column Connectors, Agilent Technologies
Fittings and Connectors
Agilent Technologies offers Ultra Inert universal press-fit column connectors.
  • Improved packaging and installation instructions
  • Ultra Inert deactivation
  • Batch certified
In the past it was necessary to use press-fit connectors with specific dimensions to connect columns of those dimensions. Modern press-fit connectors are inlaser-milled in to provide highly reproducible taper angles throughout the length of the press-fit, ensuring an excellent seal. The press-fits are now treated with Agilent Ultra Inert deactivation to provide a robust and inert flowMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable