Lab disc White Magnetic Stirrers, IKA Works
Supplier: IKA Works
0003907500
76414-092EA 798.98 CAD
76414-092
Lab disc White Magnetic Stirrers, IKA Works
Stirrers Magnetic Stirrers
Ultra-flat magnetic stirrers with magnetic coil technology. Top plate and casing are made of chemically resistant materials. Wear-free drive with no moving parts.
  • Reverses direction of rotation every 30 seconds to ensure optimum mixing (selectable)
  • Can be used at 5 to 40 °C (80% relative humidity), in an incubator or cold room
Stepless speed adjustment.
Certifications: IP protection class according to DIN EN 60529: IP 65
Order Now
SPECIFICATIONS
Environmentally Preferable