Advanced Vortex Mixers, ZX4, VELP Scientifica
Supplier: VELP SCIENTIFIC INC.
F202A0280 SA202A0280
76416-206EA 671.88 CAD
76416-206 76416-292
Advanced Vortex Mixers, ZX4, VELP Scientifica
Shakers Vortex Mixers
Vortex mixer with adjustable stirring speed, with continuous and IR sensor operating mode.
  • Variable speed operation (electronically controlled)
  • Large choice of accessories to suit tubes, plates and flasks
  • Universal voltage/frequency model
  • Accessories fit by simple pressure
  • Four large, anti-slip rubber feet for staying in required position
Optimum resistance to chemicals from technopolymer casing, excellent stability with zinc alloy base.
The infrared sensor technology ensures a drastic reduction in repetitive strain.
Certifications: IP Protection Class according to CEI EN 60259: IP 42 protection against particles and liquids.
Delivery information: Three year warranty.
Supplied with small rubber supporting plate, customizable soft foam top and stand are available as options.