RH Basic and Digital Magnetic Hotplate Strirrers, IKA
Supplier: IKA Works
0005019701 0005029701 0005019801 0004678001
76414-080EA 1299.42 CAD
76414-080 76414-082 76414-084 76414-086
RH Basic and Digital Magnetic Hotplate Strirrers, IKA
Stirrers Magnetic Hotplate Stirrers
Magnetic hotplate stirrers with stainless steel heating plate, white units have an enamelled plate. All models are easy to operate and have adjustable safety temperature from 50 to 370 °C. High heat output of 600 watts and steel-copper compound hot plate ensures rapid heating.
  • RH basic includes M 10 thread for a support rod
  • Soft-start of the stirring motor for both models
  • Strong magnetic field and wide speed range for volumes up to 15 litres
  • RH digital has digital display of temperature and speed
  • Error code display and touch keypad (RH digital)
  • Speed display scale (basic) and LED (digital)
The RH digital model has bushing according to DIN 12878 for connecting an electronic contact thermomete. Highly accurate temperature control is possible with sensor placed directly in the medium.
Maximum stirring bar length of 80 mm, Heating rate of 6 K/min.
Certifications: IP protection class according to DIN EN 60529: IP 21