Renewable Gas Purification System, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G3440-60003 G3440-60004 G3440-69003 19199C 5190-1414 G3440-80007 G3440-90000 G3440-90001
CAG3440-60003EA 1254.48 CAD
CAG3440-60003 CAAGG344060004 CAAGG344069003 CAAG19199C CAAG5190-1414 CAAGG344080007 CAAGG344090000 CAAGG344090001
Renewable Gas Purification System, Agilent Technologies
Filters Gas Filters
Renewable and long-lasting gas filter system for trapping large amounts of contaminants.
  • Highest capacity and most economical gas purification system available from Agilent
  • All cartridges are environmentally friendly and reduce waste
  • High-capacity – 850 cc or more oxygen filtration, 12 g moisture, and about 8 g hydrocarbon filtration per cartridge – in a compact footprint
  • Improves 99.995% gas purity to 99.9995% purity
The Agilent renewable gas purification system not only traps large quantities of contaminants and is long-lasting, but it is also recyclable. With average use, you’ll need a replacement cartridge once per year or after purification of about 20 gas cylinders. When you need a replacement, you can purchase a new or recycled cartridge. Recycled cartridges are refilled and certified to the full specification of new cartridges.