Flame Photometric Detector (FPD) for GC, Agilent Technologies
Supplier: Agilent
19256-20690 1000-1438 19256-20910 19256-60700 5188-5245 G3436-67000 19256-80045 5061-5886 19256-80045 G1535-60600 19256-80030 G1535-60600 19256-60800 1000-1437 19256-60800 19256-20690 5061-5890 19256-80010 19256-60700 G3436-67000 5061-5891 19256-80010 5080-8846 5061-5886 19256-80030 19256-20910 5188-5953 5061-5890
CAAGG343667000EA 1032.9 CAD
CAAGG343667000 AG19256-20910 AGG1535-60600 AG5061-5886 CAAG1925660700 CAAG1925660800 CAAG1925680010 CAAG5061-5886 CAAG1925620690 AG19256-20690 CAAG1925620910 AG19256-80045 CA19256-80030 CAAGG153560600 AG19256-80030 AG19256-80010 101471-128 AG19256-60700 AG19256-60800 CAAG1925680045 AGG3436-67000 101471-908 CAAG5061-5890 101472-004 AG5061-5891 AG5080-8846 101471-130 AG5061-5890
Flame Photometric Detector (FPD) for GC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
In 2005, Agilent released an improved FPD with a design based on a one-piece, deactivated transferline jet assembly and improved optics.
  • The FPD uses three gases: air and hydrogen to support the flame, and nitrogen makeup for capillary columns
  • The flow rates are critical for optimizing performance
In 2005, Agilent released an improved FPD with minimum detectable levels (MDL) of 3.6 pg/s for sulfur and 60 fg/s for phosphorus. This is more than a 5 times improvement for sulfur. The updated design is based on a one-piece, deactivated transferline jet assembly and improved optics. Upgrade kits are available.