Heratherm® Drying Ovens, General Protocol Series, Thermo Scientific
Supplier: Thermo Fisher Scientific
51028112 51028872 51028874 51028121 51028873 51028875
76416-326EA 4534.6 CAD
76416-326 76416-328 76416-330 76416-332 76416-334 76416-336
Heratherm® Drying Ovens, General Protocol Series, Thermo Scientific
Ovens Standard Ovens
Heratherm® General Protocol ovens which provide an ideal heating and drying solution for research, clinical or industrial needs.
  • Operating temperatures from 50 to 250 °C
  • Automatic over-temperature alarm system
  • Low overall energy consumption
  • Built in timer
  • Large, easy to read vacuum fluorescent display
  • Doors can be opened at over 180 °C
Easy to use, microprocessor-controlled touch button operation. Corrosion-resistant stainless steel chambers with rounded corners.
Delivery information: Two year warranty.