Bond Elut PBA Plates
Supplier: Agilent
Bond Elut
7541201C A396121C 7541201C
AG7541201CEA 1366.73 CAD
AG7541201C 101402-242 101473-730
Bond Elut PBA Plates
Microplates Solid Phase Extraction Plates (SPE)
Unique bond Elut PBA silica sorbent uses phenylboronic acid functionality to retain analytes via a reversible covalent bond, with high specificity and cleanliness.
  • Unique phenylboronic acid sorbent
  • High specificity for cis-diol compounds
  • Amenable for a broad range of biomolecule applications
Bond Elut PBA is a unique silica SPE sorbent containing a phenylboronic acid functionality that can retain analytes through a reversible covalent bond. This very strong covalent retention mechanism enables high specificity and cleanliness.
The boronate group has a strong affinity for cis-diol containing compounds, such as catechols, nucleic acids, some proteins, carbohydrates, and PEG compounds. Amino alcohols, alpha-hydroxy amides, keto compounds, and others can also be retained.