Bond Elut NH₂ Plates
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
7540501C 7540502C
AG7540501CEA 810.54 CAD
AG7540501C AG7540502C AG75405050
Bond Elut NH₂ Plates
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut NH₂ is a weak anion exchanger and is a good choice for retention of very strong anions that may be irreversibly retained on other anion exchange sorbents.
  • Normal phase or anion-exchange sorbent
  • Weaker anion exchange than SAX
  • Amenable to separating structural isomers
Bond Elut NH₂ is a weaker anion exchanger than sorbents such as SAX (a quaternary amine sorbent that is always charged), and is therefore a better choice for retention of very strong anions, such as sulfonic acids, which may retain irreversibly on a SAX sorbent. Similar to diol an SI sorbents, bond elut NH₂ is excellent for the separation of structural isomers.