Bond Elut CBA Plates
Supplier: Agilent
Bond Elut
A496061C A3960625 7540601C A396061C 7540601C A4960625 75406050
AG75406050EA 790.85 CAD
AG75406050 101402-216 101402-214 101473-714 101473-822 101473-820 AG7540601C
Bond Elut CBA Plates
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut CBA (Carboxylic acid) is a mid-polarity sorbent and weak cation exchanger that is a good choice for working with strong cations.
  • Cation exchange with no need for extreme, basic conditions
  • Wider selectivity range provides more eluent options
  • Polar or nonpolar depending on matrix or solvent
CBA is a midpolarity sorbent and weak cation exchanger (pKa 4.8). It can be used with a wider range of counterions than lower pKa sorbents like SCX, and will demonstrate easier elution of quaternary amine functionalized analytes.