Intuvo Flow Chips, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G4581-60031 G4590-60009 G4581-60032 G4581-60033 G4583-60331 G4583-60333 G4583-60335 G4583-60336 G4583-60621 G4588-60302 G4588-60322 G4588-60402 G4588-60502 G4588-60601 G4588-60701 G4588-60721 G4590-60109
CAG4581-60031EA 721.36 CAD
CAG4581-60031 CAG4590-60009 CAAGG458160032 CAAGG458160033 CAAGG458360331 CAAGG458360333 CAAGG458360335 CAAGG458360336 CAAGG458360621 CAAGG458860302 CAAGG458860322 CAAGG458860402 CAAGG458860502 CAAGG458860601 CAAGG458860701 CAAGG458860721 CAAGG459060109
Intuvo Flow Chips, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Intuvo Flow Chips are modular, microfluidic components that create connections between the inlet, column, and detector(s) without the need for ferrules, and can be easily swapped in minutes.
  • Highly inert fused silica Flow Chips enable the connections you need, fast
  • Easily installed in minutes
  • No more guesswork andndash
  • automatic system configuration via Smart Keys
  • Simplifies capillary flow techniques like backflush or splitting to dual detectors
Intuvo Flow Chips are modular, microfluidic components that create connections between the inlet, column,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable