CrossLab Metering Device Supplies for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
0905-1599 5022-2175 5062-8587 5064-8293 5063-6586 5063-6589 0905-1294 0905-1717 5068-0198 G5611-21503 5067-4695
CA0905-1599EA 93.05 CAD
CA0905-1599 CA5022-2175 CA5062-8587 CA5064-8293 CA76508-064 CA76508-066 CA76514-530 CA0905-1717 CA5068-0198 CAG5611-21503 CA5067-4695
CrossLab Metering Device Supplies for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Designed to prevent loss in injection volume precision or metering device leaking.
Agilent LC Metering Device Supplies are designed to operate with your LC instruments to work together seamlessly for maximum efficiency and performance–regardless of application area. Supporting your efforts to increase your analytical, instrument, and laboratory efficiency in your everyday work.
Agilent replacement metering device supplies for HPLC, including the metering device seal and piston, are designed to prevent loss in injection volume precision or metering device leaking.