Capillary Flow Technology Supplies, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G2855-20555 G2855-20530 G2855-20590 G2855-60200 G2855-60570 G2855-20532 G3184-60065
CAG2855-20555EA 56.46 CAD
CAG2855-20555 CA76515-264 CAG2855-20590 CAG2855-60200 CAG2855-60570 CAAGG285520532 CAAGG318460065
Capillary Flow Technology Supplies, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Designed for Capillary Flow devices Agilent offers GC accessories based on our proprietary Capillary Flow Technology. These accessories increase system productivity and performance.
  • Deans Switch device supplies the analysis of complex samples
  • Purged Effluent Splitter for inert, leak-free column effluent splitting
For leak-free, low dead volume and inert column connections with capillary flow accessories, such as the Deans Switch or QuickSwap MS Interface, use only Siltite ferrules and specified nuts. For Capillary Flow devices, use deactivated fused silica tubing. Do not use tubing that has been coated with stationary phase.