Bond Elut LMS Plates
Supplier: Agilent
Bond Elut
7541002C 7541002C A4961010 A3961010
AG7541002CEA 899.94 CAD
101402-238 AG7541002C 101473-726 101473-836
Bond Elut LMS Plates
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut LMS (Large Molecule Size) enables elution without the secondary reactions of additives.
  • Ultraclean styrene-divinylbenzene polymer
  • Optimized 75 m particle size for reproducible flow
  • High capacity and surface area for efficient extraction
Bond Elut LMS polymeric sorbent lets you elute without having to add amine modifiers, buffers, or acids. The elimination of secondary interactions means that elution of analytes can be achieved with pure organic solvents or HPLC mobile phase-compatible solvent mixtures of low ionic strength. These characteristics are crucial to allow compatibility with LC/MS or other delicate analytical techniques.