Bond Elut™ Adapters and Caps, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
12131001 12131003
CA12131001PK 137.99 CAD
CA12131001 CAAG12131003
Bond Elut™ Adapters and Caps, Agilent Technologies
Fittings and Connectors
Bond Elut adapters and caps connect series of SPE cartridges and produce positive pressure seals.
  • Adapters connect SPE cartridges in series for large samples
  • Adapters expand cartridge volume for even more applications
  • Adapters allow you to transfer large-volume samples to any SPE cartridge
  • Caps convert 1, 3 and 6 ml cartridges for use in Gilson ASPEC SPE systems
  • Caps enhance the high throughput compatibility of Bond Elut cartridges
  • Specially engineered for leak-free operation
Bond Elut adapters fit on top of any Bond Elut cartridge and contain aMore Product Information