Podium Lectern, Diversified Woodcrafts

Supplier: Diversified Woodcrafts
IP-K IP-M IPTD-K IPTD-M
470328-706EA 1532.6 CAD
470328-706 470328-708 470328-710 470328-712
Podium Lectern, Diversified Woodcrafts
Lecterns
Finally a lectern to match the rest of the furniture in your skilled spaces.

  • Large 25W×22"D worksurface
  • Overall Size: 49"H (student side) 43"H (instructor side)
  • Your choice of maple or Oak veneer construction
  • Three fixed shelves for storing supplies
  • Units roll easily on 2" casters
  • Worksurface has a pencil ledge and rests at a comfortable 15 degree angle
  • Your choice to order with 12 Testing Dividers loaded in the base
  • Option of ordering bracket for hanging Testing Dividers

FinaThis lectern is constructed of your choice of maple or Oak veneers with andMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR