Sartoclear Dynamics® Lab V Clarification and Sterile Filtration Kits, Sartorius
Supplier: Sartorius
SDLV005001E02 SDLV005001F02 SDLV005002E02
76420-782PK 645.78 CAD
76420-782 76420-784 76420-786 76420-788 76420-790 76420-792 76420-794 76420-796 76420-798 76420-800 76420-802 76420-804 76420-806 76420-808
Sartoclear Dynamics® Lab V Clarification and Sterile Filtration Kits, Sartorius
Filters Bottle Top Filters
Sartoclear Dynamics® Lab products are filtration kits designed for the clarification and sterile filtration of up to 1000 ml mammalian cell cultures.
  • Ready to use
  • Fast and effortless
  • Clarification technology available for laboratories but also for process volumes
Sartoclear Dynamics® Lab kits enable clarification and sterile filtration to be performed in a single step. These kits simplify the process by fully eliminating the centrifugation step otherwise needed for clarification. As a result, they enable harvested cell cultures to be efficientlyMore Product Information