Column Installation Pre-Swaging Tools for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
19251-80680 G3440-80217 G3440-80227 G3440-80218 G3440-80247
CA19251-80680EA 127.01 CAD
CA19251-80680 CAG3440-80217 CAG3440-80227 CAAGG344080218 CAAGG344080247
Column Installation Pre-Swaging Tools for GC, Agilent Technologies
Puller and Inserter Tools
Handy stainless-steel tools for proper column installation lengths into the most popular GC inlets.
  • Available for Agilent 7890/6890 model split/split less, multimode, and purged packed inlets
  • Each tool has a fitting specific to its ferrule type to provide optimal sealing
  • one for graphite ferrules, one for metal ferrules
  • Easy to read markings for split/split less, multimode, and purged packed inlet lengths
  • A ferrule release hole on the preswaging tool for metal ferrules (G3440-80218) ensures against overtightening resulting in a stuck ferrule