Bond Elut PH Plates
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
7541501C 7541502C
AG7541501CEA 810.54 CAD
AG7541501C AG7541502C
Bond Elut PH Plates
Microplates Solid Phase Extraction Plates (SPE)
Bond Elut PH phenyl sorbent is a moderately non-polar sorbent commonly used for non-polar extractions, useful for retention of planar, conjugated organic molecules.
  • Added selectivity compared to other nonpolar sorbents
  • Enhanced retention of planar, conjugated organic molecules
  • Similar polarity to C8
Bond Elut PH is a nonpolar bonded silica material, which exhibits a different selectivity to alkyl or aliphatic functionalized phases, such as C8 or cyclohexyl. The electron density present in the aromatic ring affords an enhancement in the retention ofMore Product Information