Sartolab® RF 50 Tube Top Vacuum Filtration Systems
Supplier: Sartorius
180E012 180F012 180E01---------2 180F01---------2 180E01E8
76420-776PK 384.1 CAD
76420-776 76420-778 CA76894-062 CA76894-064 CA77578-296
Sartolab® RF 50 Tube Top Vacuum Filtration Systems
Filters Bottle Top Filters
Sartolab® RF 50 vacuum filtration units are single-use units for the filtration of sample volumes up to 50 ml. They are available with either a 0.22 or 0.45 µm polyethersulfone membrane.
  • Highest flow rates with an asymmetric membrane and a large filtration surface
  • No loss of protein with a low protein affinity membrane
  • Low dead volume due to an optimized membrane support
  • Minimized risk of contamination with a complete ready-to-use unit (no additional transfer of liquid necessary for storage)
The design of the yellow adapter connecting the funnel to the conical tube ensures a vacuum-tight seal and enables the filtration unit to be used on the Sartolab® MultiStation for filtration of up to 6 samples in parallel with one vacuum source. However, Sartolab® RF 50 can also be used alone by directly connecting the unit’s tubing connector to your vacuum source.
Both membranes provide fast flow rates and ensure low protein binding and extractables. Both versions are individually-packed, sterile, and ready to use. The 0.22 µm version is ideal for sterile filtration of cell cultures, buffers and aqueous solutions, while the 0.45 µm version is ideal for clarification.
Pressure: –350 to –700 mbar.
Delivery information: The Sartolab® RF 50 vacuum filtration unit comprises a funnel with dust cover, a 50 ml conical tube with graduation and writing field, a tubing connector for vacuum connection and a screw cap for storage of your filtrate (individually wrapped to maintain sterility).