FlaskScrubber® Vantage® Glassware Washers, Labconco®
Supplier: Labconco
422101010 422101013
76420-732EA 40290 CAD
76420-732 76420-734
FlaskScrubber® Vantage® Glassware Washers, Labconco®
Glassware Washers
Versatile and efficient glassware washers designed for efficient and consistent washing of narrow-neck glassware and general labware in critical washing environments.
  • Touch screen display with intuitive CleanWorks OS software
  • Flexible design allows for use of spindle injection and standard racks
  • Maximum wash temperature 93 °C removes tough contaminants
  • Conductivity sensor and HEPA-filtered forced air spindle drying with SpeedVent technology
  • Durable Type 304 stainless steel chamber and racks and detergent storage cabinet
  • 36-place Spindle Injection Rack included
  • Liquid detergent and acid neutralizing rinse aid pumps included
These glassware washers are designed for efficient and consistent washing of narrow-neck glassware and general purpose labware in critical environments such as pharmaceutical laboratories. The CleanWorks OS software is accessible through a full-color touch screen interface and provides intuitive and clear monitoring of wash loads – ensuring repeatable, effective and traceable cleaning. Vantage models reach a maximum of 93 °C wash temperature and monitor water cleanliness with a standard conductivity sensor.
FlaskScrubber Vantage models utilize HEPA filtered forced air spindle injection and SpeedVent heat-management technology to quickly dry labware. All Vantage models include a 36-place lower spindle rack, dispensing pumps for liquid detergent and acid neutralizing rinse, and a batch viewing window. Vantage models may be outfitted with a wide variety inserts and additional racks to accommodate all types of labware.
Certifications: ETL, cETL, and CE listed. Compliant with UL61010, CAN/CSA C22.2, CE.
Ordering information: Two year warranty on parts and labor. Optional accessories include various inserts for specific glassware types, detergents, spindle and standard racks, labware carts, and water purifiers.
Delivery information: Lower spindle rack included.
Caution: All single phase models require hard wiring to a 20 amp dedicated circuit. All three phase models require hard wiring to a 30 amp dedicated circuit.