Headspace Vials and Caps, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5182-0840 5182-0837 5182-0839 5188-2753 5188-5392 5182-0838 5183-4474 5183-4475 5190-2285 5190-2286 5190-2288 8010-0412 8010-0413 5185-5957 5190-2238 5190-2287 5190-9598 8010-0152 8010-0179 5067-0226 5067-0227 5188-6537 5188-6538 5190-2239 8010-0180 5191-5634
CA5182-0840PK 194.79 CAD
CA5182-0840 CA76508-134 CA76508-136 CA76508-180 CA76508-190 CA76514-794 CA76514-862 CA76514-864 CA5190-2285 CA5190-2286 CA5190-2288 CA8010-0412 CA8010-0413 CAAG5185-5957 CAAG5190-2238 CAAG5190-2287 CAAG5190-9598 CAAG8010-0152 CAAG8010-0179 CA5067-0226 CA5067-0227 CA5188-6537 CA5188-6538 CA5190-2239 CA8010-0180 CAAG5191-5634
Headspace Vials and Caps, Agilent Technologies
Vials
Engineered to prevent sample contamination, with caps that provide consistently secure seals.
  • Beveled top for maximum seal security
  • Available in flat bottom glass vial or rounded bottom glass vial designs
  • Optional pressure safety release cap at 45 psi
  • High-performance cap septa designed for higher temperature usage up to 300 °C
Crimp caps are most common however many customers use screw sample and application dependent. Headspace vials come in both flat bottom and round bottom styles for use in a wide variety of headspace instrument platforms.More Product Information