Vivaflow® 200 Modular Tangential Flow System, Sartorius
Supplier: Sartorius
Vivaflow®
VFS204 VFS204
CA76408-736EA 4159.66 CAD
76408-736 CA76408-736
Vivaflow® 200 Modular Tangential Flow System, Sartorius
Filters Tangential Flow Systems
Vivaflow® 200 is a ready to use and reusable laboratory crossflow cassette for concentrating up to 5 L samples.
  • Optimal for cell culture supernatant and virus concentration
  • High flow rate and recovery
The membrane surface per device is 200 cm². One unit comes with all necessary accessories for running the device with a laboratory pump and a size 16 pump head.
For speeding up the concentration, two cassettes can be run simultaneously.
Packaging: Pump, easy load pump head (size 16), tubing, 500 ml sample/diafiltration reservoir.