Vivaspin® 2 Centrifugal Filtration Units, Sartorius
Supplier: Sartorius
VS0201 VS0202 VS0211 VS0212 VS0221 VS0222 VS0231 VS0232 VS0241 VS0242 VS0291 VS0292 VS02H01 VS02H02 VS02H11 VS02H12 VS02H21 VS02H22 VS02H91 VS02H92 VS02V2 VS02X2 VS02V1
CA76408-900EA 239.65 CAD
CA76408-900 CA76408-902 CA76408-904 CA76408-906 CA76408-908 CA76408-910 CA76408-912 CA76408-914 CA76408-916 CA76408-918 CA76408-920 CA76408-922 CA76408-924 CA76408-926 CA76408-928 CA76408-930 CA76408-932 CA76408-934 CA76408-936 CA76408-938 CA76408-940 CA76408-942 CA76408-944 CA76408-946 CA76408-948 CA76408-950 CA76408-952 CA76408-958 CA76408-960 CA76408-962 CA76408-964 CA76408-959
Vivaspin® 2 Centrifugal Filtration Units, Sartorius
Filters Centrifugal Filters
Vivaspin® concentrators are disposable ultrafiltration devices for the concentration of biological samples of 0.4 to 2 ml.
  • High retentate recovery >90%
  • Vertical membrane design for ultra-fast concentration
  • Safe sample concentration due to dead-stop volume
  • Convenient concentrate recovery by pipetting
  • Volume graduation for easy reading/monitoring
Vivaspin® 2 can effectively be used in swing bucket or fixed angle rotors accepting 15 ml centrifuge tubes. The patented vertical membrane design and thin channel filtration chamber (US 5647990) minimizes membrane fouling and provides high speed concentrations, even with particle-laden solutions.
Vivaspin® 2 combines the speed of the classic Vivaspin® products with low internal surface and membrane area in order to achieve superior recoveries from very dilute solutions.